missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine CD8A / MAL Protein

A DNA sequence encoding the Bos taurus (Bovine) CD8A / MAL, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5800-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15937859

Quantità Prezzo
1 309.00€ / 10µg
6 288.51€ / 10µg
Spedizione stimata
18-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_776440.1
CD8A / MAL Protein
10 μg
Not determined.
Extracellular Domain
Bovine
His-SUMO
Tris-based buffer, 50% glycerol
281060
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD8A
Recombinant Protein
E. coli
LSFRMSPTQK ETRLGEKVEL QCELLQSGMA TGCSWLRHIP GDDPRPTFLM YLSAQRVKLA EGLDPRHISG AKVSGTKFQL TLSSFLQEDQ GYYFCSVVSN SILYFSNFVP VFLPAKPATT PAMRPSSAAP TSAPQTRSVS PRSEVCRTSA GSAVDTSRLD FACN
Videos
SDS
Documenti