missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine MTHFD2 Protein
A DNA sequence encoding the Bos taurus (Bovine) MTHFD2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7520-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001069223.1 | |
MTHFD2 Protein | |
Research Use Only | |
MTHFD2 | |
Recombinant Protein | |
E. coli | |
EAVVISGRKL AEQIKQEVRQ EVEEWVASGN KRPHLSVVLV GENPASQSYV LNKTRAAASV GINSETILKP ASISEEELLN LINKLNNDDN VDGLLVQLPL PEHIDERKVC NAVSPDKDVD GFHVINVGRM CLDQCSMLPA TPWGVWEIIK RTGIPTLGKN VVVAGRSKNV GMPIAMLLHT DGAHERPGGD ATVTISHRYT PKEELKKHTA LADIVISAAG IPNLITADMI KEGAAVIDVG INRIQDPITA KPKLVGDVDF EGVKKKAGYI TPVPGGVGPM TVAMLMKNTI IAAKKVLRLE EQEVLKSKEL GVASN | |
Greater than 90% as determined by SDS-PAGE. |
517539 | |
500 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |