missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum clpP Protein

A DNA sequence encoding the Clostridium botulinum (strain Hall / ATCC 3502 / NCTC 13319 / Type A) clpP, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7429-ec-50ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16058830

Quantità Prezzo
1 437.00€ / 50µg
6 407.98€ / 50µg
Spedizione stimata
20-10-2020
Aggiungi al carrello


Specifica

Specifica

WP_003357557.1
clpP Protein
50 μg
Not determined.
Full Length
C. botulinum
His-SUMO
Tris-based buffer, 50% glycerol
5187372
Greater than 90% as determined by SDS-PAGE.
Research Use Only
clpP
Recombinant Protein
E. coli
MSLVPVVVEQ TNRGERSYDI YSRLLKDRII MLSEEVNDTT ASLIVAQLLF LEAEDPDKDI HLYINSPGGS ITSGMAIYDT MQYIKPDVST ICVGMAASMG AFLLAAGAKG KRYALPNSEV MIHQPLGGFR GQATDIGIHA ERILKMKKKL NTILSDRTGK PLEQVELDTE RDHFLSAEEA KEYGLIDEVI DKKK
Videos
Documenti