missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia W4 Protein
A cDNA sequence encoding the Chlamydia W4 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11416-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
W4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/1ml) 20mM Tris-HCl, pH 7.2, 50% glycerol and 1.5M urea. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chlamydia | |
Untagged | |
Protein is >90% pure as determined by SDS PAGE. |