missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Envelope-2 & 4 Protein

A cDNA sequence encoding the Dengue Envelope-2 & 4 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11627-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15937848

Quantità Prezzo
1 1462.00€ / 1mg
3 1375.16€ / 1mg
Spedizione stimata
18-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

Dengue Envelope-2 & 4 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
The protein solution contains Phosphate buffered saline, pH 7.4.
Videos
SDS
Documenti