missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog CMA1 / Chymase 1 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5843-ec-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15938179

Quantità Prezzo
1 1245.00€ / 200µg
3 1160.96€ / 200µg
Spedizione stimata
11-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_001013442.1
CMA1 / Chymase 1 Protein
200 μg
Not determined.
Full Length
Dog
His-SUMO
Tris-based buffer, 50% glycerol
490628
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CMA1
Recombinant Protein
E. coli
IIGGTESKPH SRPYMAHLEI LTLRNHLASC GGFLIRRNFV LTAAHCAGRF IMVTLGAHNI QKKEDTWQKL EVIKQFPHPK YDDLTLRHDI MLLKLKEKAN LTLAVGTLPL SPQFNFVPPG RMCRVAGWGK RQVNGSGSDT LQEVKLRLMD PQACRHYMAF DHNLQLCVGN PRKTKSAFKG DSGGPLLCAG VAQGIVSYGQ NDAKPPAVFT RISHYRPWIN KVLKQNKA
Videos
SDS
Documenti