missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog MMP-9 / CLG4B Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) MMP-9 / CLG4B, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9280-ye-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16057991

  • 1100.00€ / 100µg
Spedizione stimata: 28-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Specifica

Specifica

MMP-9 / CLG4B Protein
Research Use Only
MMP9
Recombinant Protein
Yeast
FQTFEGDLKW HHNDITYWIQ NYSEDLPRDV IDDAFARAFA VWSAVTPLTF TRVYGPEADI IIQFGVREHG DGYPFDGKNG LLAHAFPPGP GIQGDAHFDD EELWTLGKGV VVPTHFGNAD GAPCHFPFTF EGRSYSACTT DGRSDDTPWC STTADYDTDR RFGFCPSEKL YAQDGNGDGK PCVFPFTFEG RSYSTCTTDG RSDGYRWCST TADYDQDKLY GFCPTRVDSA VTGGNSAGEP CVFPFIFLGK QYSTCTREGR GDGHLWCATT SNFDRDKKWG FCPDQGYSLF LVAAHEFGHA LGLDHSSVPE ALMYPMYSFT EGPPLHEDDV RGIQHLYGPR PEPEPQPPTA PPTAPPTVCA TGPPTTRPSE RPTAGPTGPP AAGPTGPPTA GPSEAPTVPV DPAEDICKVN IFDAIAEIRN YLHFFKEGKY WRFSKGKGRR VQGPFLSPST WPALPRKLDS AFEDGLTKKT FFFSGRQVWV YTGTSVVGPR RLDKLGLGPE VTQVTGALPQ GGGKVLLFSR QRFWSFDVKT QTVDPRSAGS VEQMYPGVPL NTHDIFQYQE KAYFCQDRFY WRVNSRNEVN QVDEVGYVTF DILQCPED
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali