missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Lipid kinase YegS Protein
A DNA sequence encoding the Escherichia coli (strain K12) Lipid kinase YegS, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6965-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_416590.1 | |
Lipid kinase YegS Protein | |
Research Use Only | |
yegS | |
Recombinant Protein | |
E. coli | |
MAEFPASLLI LNGKSTDNLP LREAIMLLRE EGMTIHVRVT WEKGDAARYV EEARKFGVAT VIAGGGDGTI NEVSTALIQC EGDDIPALGI LPLGTANDFA TSVGIPEALD KALKLAIAGD AIAIDMAQVN KQTCFINMAT GGFGTRITTE TPEKLKAALG SVSYIIHGLM RMDTLQPDRC EIRGENFHWQ GDALVIGIGN GRQAGGGQQL CPNALINDGL LQLRIFTGDE ILPALVSTLK SDEDNPNIIE GASSWFDIQA PHDITFNLDG EPLSGQNFHI EILPAALRCR LPPDCPLLR | |
Greater than 90% as determined by SDS-PAGE. |
946626 | |
10 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |