missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli LptA Protein

A DNA sequence encoding the Escherichia coli (strain K12) LptA, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7331-ec-10ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16058250

Quantità Prezzo
1 309.00€ / 10µg
6 288.51€ / 10µg
Spedizione stimata
23-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_417667.1
LptA Protein
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
947920
Greater than 90% as determined by SDS-PAGE.
Research Use Only
lptA
Recombinant Protein
E. coli
VTGDTDQPIH IESDQQSLDM QGNVVTFTGN VIVTQGTIKI NADKVVVTRP GGEQGKEVID GYGKPATFYQ MQDNGKPVEG HASQMHYELA KDFVVLTGNA YLQQVDSNIK GDKITYLVKE QKMQAFSDKG KRVTTVLVPS QLQDKNNKGQ TPAQKKGN
Videos
Documenti