missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli NFNB Protein
A cDNA sequence encoding the NFNB was constructed and used to recombinantly synthesize the protein.
120.00€ - 3595.00€
Specifica
Nome | NFNB Protein |
---|---|
Purezza | Greater than 95.0% as determined by SDS-PAGE. |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15941709
|
enQuireBio™
QP12838-5UG |
5 μg |
120.00€
5µg |
Eseguire il login per visualizzare lo stock disponibile |
15931709
|
enQuireBio™
QP12838-20UG |
20 μg |
181.00€
20µg |
Eseguire il login per visualizzare lo stock disponibile |
15921709
|
enQuireBio™
QP12838-1MG |
1 mg |
3595.00€
1mg |
Eseguire il login per visualizzare lo stock disponibile |
Specifica
NFNB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDIISVALKR HSTKAFDASK KLTPEQAEQI KTLLQYSPSS TNSQPWHFIV ASTEEGKARV AKSAAGNYVF NERKMLDASH VVVFCAKTAM DDVWLKLVVD QEDADGRFAT PEAKAANDKG RKFFADMHRK DLHDDAEWMA KQVYLNVGNF LLGVAALGLD AVPIEGFDAA ILDAEFGLKE KGYTSLVVVP VGHHSVEDFN ATLPKSRLPQ NITLTEV |
Greater than 95.0% as determined by SDS-PAGE. | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
1 mg/ml solution containing 20mM Tris pH 8.0, 1mM DTT, 0.05M NaCl and 10% glycerol. |