missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli S-ribosylhomocysteine lyase Protein
A DNA sequence encoding the Escherichia coli (strain K12) S-ribosylhomocysteine lyase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7508-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_417172.1 | |
S-ribosylhomocysteine lyase Protein | |
Research Use Only | |
luxS | |
Recombinant Protein | |
E. coli | |
PLLDSFTVDH TRMEAPAVRV AKTMNTPHGD AITVFDLRFC VPNKEVMPER GIHTLEHLFA GFMRNHLNGN GVEIIDISPM GCRTGFYMSL IGTPDEQRVA DAWKAAMEDV LKVQDQNQIP ELNVYQCGTY QMHSLQEAQD IARSILERDV RINSNEELAL PKEKLQELHI | |
Greater than 90% as determined by SDS-PAGE. |
947168 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |