missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS3 Protein
A cDNA sequence encoding the HCV NS3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12162-B-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hepatitis C HCV NS3 Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Biotin |
HCV NS3 Biotin labeled protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
8M urea, 20mM Tris-HCl pH 8.0, 10mM b-mercaptoethanol. |