missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-3b Protein

A cDNA sequence encoding the HCV NS5 Genotype-3b was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP12194-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15939658

Quantità Prezzo
1 733.00€ / 500µg
5 688.36€ / 500µg
Spedizione stimata
04-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

Hepatitis C HCV NS5 Genotype-3b Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
GST
HCV NS5 Genotype-3b protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl, pH 8.0, 0.2% Triton-X and 50% Glycerol.
Videos
SDS
Documenti