missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant hepatitis Non-structural polyprotein pORF1 Protein
A DNA sequence encoding the Hepatitis E virus genotype 1 (isolate Human/Pakistan/Sar-55) (HEV-1) Non-structural polyprotein pORF1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7125-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
hepatitis Non-structural polyprotein pORF1 Protein | |
Research Use Only | |
ORF1 | |
Recombinant Protein | |
E. coli | |
EVFWNHPIQR VIHNELELYC RARSGRCLEI GAHPRSINDN PNVVHRCFLR PAGRDVQRWY TAPTRGPAAN CRRSALRGLP AADRTYCFDG FSGCNFPAET GIALYSLHDM SPSDVAEAMF RHGMTRLYAA LHLPPEVLLP PGTYRTASYL LIHDGRRVVV TYEGDTSAGY NHDVSNLRSW I | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |