missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HSV-2 gG Protein
A cDNA sequence encoding the HSV-2 gG was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12346-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
HSV HSV-2 gG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HSV-2 gG solution contains 25mM Tris-HCl pH 8.0. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
Untagged | |
Protein is >99% pure as determined by Analytical HPLC and Mass Spec. |