missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S25 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S25, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8329-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001019.1 | |
40S ribosomal protein S25 Protein | |
Research Use Only | |
RPS25 | |
Recombinant Protein | |
E. coli | |
MPPKDDKKKK DAGKSAKKDK DPVNKSGGKA KKKKWSKGKV RDKLNNLVLF DKATYDKLCK EVPNYKLITP AVVSERLKIR GSLARAALQE LLSKGLIKLV SKHRAQVIYT RNTKGGDAPA AGEDA | |
Greater than 90% as determined by SDS-PAGE. |
6230 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |