missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L30 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L30, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6620-ec-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16064600

Quantità Prezzo
1 1855.00€ / 1mg
3 1731.09€ / 1mg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_000980.1
60S ribosomal protein L30 Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6156
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RPL30
Recombinant Protein
E. coli
MVAAKKTKKS LESINSRLQL VMKSGKYVLG YKQTLKMIRQ GKAKLVILAN NCPALRKSEI EYYAMLAKTG VHHYSGNNIE LGTACGKYYR VCTLAIIDPG DSDIIRSMPE QTGEK
Videos
Documenti