missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L8 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8326-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000964.1 | |
60S ribosomal protein L8 Protein | |
Research Use Only | |
RPL8 | |
Recombinant Protein | |
E. coli | |
RVIRGQRKGA GSVFRAHVKH RKGAARLRAV DFAERHGYIK GIVKDIIHDP GRGAPLAKVV FRDPYRFKKR TELFIAAEGI HTGQFVYCGK KAQLNIGNVL PVGTMPEGTI VCCLEEKPGD RGKLARASGN YATVISHNPE TKKTRVKLPS GSKKVISSAN RAVVGVVAGG GRIDKPILKA GRAYHKYKAK RNCWPRVRGV AMNPVEHPFG GGNHQHIGKP STIRRDAPAG RKVGLIAARR TGRLRGTKTV QEKEN | |
Greater than 90% as determined by SDS-PAGE. |
6132 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |