missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Actin-like protein 7A Protein

A DNA sequence encoding the Homo sapiens (Human) Actin-like protein 7A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8283-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16013761

Quantità Prezzo
1 434.28€ / 100µg
6 408.22€ / 100µg
Spedizione stimata
13-05-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_006678.1
Actin-like protein 7A Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
10881
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ACTL7A
Recombinant Protein
E. coli
MWAPPAAIMG DGPTKKVGNQ APLQTQALQT ASLRDGPAKR AVWVRHTSSE PQEPTESKAA KERPKQEVTK AVVVDLGTGY CKCGFAGLPR PTHKISTTVG KPYMETAKTG DNRKETFVGQ ELNNTNVHLK LVNPLRHGII VDWDTVQDIW EYLFRQEMKI APEEHAVLVS DPPLSPHTNR EKYAEMLFEA FNTPAMHIAY QSRLSMYSYG RTSGLVVEVG HGVSYVVPIY EGYPLPSITG RLDYAGSDLT AYLLGLLNSA GNEFTQDQMG IVEDIKKKCC FVALDPIEEK KVPLSEHTIR YVLPDGKEIQ LCQERFLCSE MFFKPSLIKS MQLGLHTQTV SCLNKCDIAL KRDLMGNILL CGGSTMLSGF PNRLQKELSS MCPNDTPQVN VLPERDSAVW TGGSILASLQ GFQPLWVHRF EYEEHGPFFL YRRCF
Videos
SDS
Documenti