missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACY1 / Aminoacylase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) ACY1 / Aminoacylase-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8038-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000657.1 | |
ACY1 / Aminoacylase-1 Protein | |
Research Use Only | |
ACY1 | |
Recombinant Protein | |
E. coli | |
MTSKGPEEEH PSVTLFRQYL RIRTVQPKPD YGAAVAFFEE TARQLGLGCQ KVEVAPGYVV TVLTWPGTNP TLSSILLNSH TDVVPVFKEH WSHDPFEAFK DSEGYIYARG AQDMKCVSIQ YLEAVRRLKV EGHRFPRTIH MTFVPDEEVG GHQGMELFVQ RPEFHALRAG FALDEGIANP TDAFTVFYSE RSPWWVRVTS TGRPGHASRF MEDTAAEKLH KVVNSILAFR EKEWQRLQSN PHLKEGSVTS VNLTKLEGGV AYNVIPATMS ASFDFRVAPD VDFKAFEEQL QSWCQAAGEG VTLEFAQKWM HPQVTPTDDS NPWWAAFSRV CKDMNLTLEP EIMPAATDNR YIRAVGVPAL GFSPMNRTPV LLHDHDERLH EAVFLRGVDI YTRLLPALAS VPALPSDS | |
Greater than 80% as determined by SDS-PAGE. |
95 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |