missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AIMP1 / EMAP2 / SCYE1 Protein

A DNA sequence encoding the Human AIMP1 / EMAP2 / SCYE1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10366-ec-5ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943638

Quantità Prezzo
1 105.00€ / 5µg
10 98.00€ / 5µg
Spedizione stimata
04-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_001135887.1
AIMP1 / EMAP2 / SCYE1 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SKPIDVSRLD LRIGCIITAR KHPDADSLYV EEVDVGEIAP RTVVSGLVNH VPLEQMQNRM VILLCNLKPA KMRGVLSQAM VMCASSPEKI EILAPPNGSV PGDRITFDAF PGEPDKELNP KKKIWEQIQP DLHTNDECVA TYKGVPFEVK GKGVCRAQTM SNSGIK
9255
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
AIMP1
The ED50 as determined by the apoptotic effect using serum free human MCF-7 cells is less than 40 ng/ml, corresponding to a specific activity of > 2.5 M 104 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documenti