missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human APPBP1/UBA3 Complex (NEDD8 E1) Protein, CF
Description
Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human APPBP1/UBA3 Complex (NEDD8 E1) Protein concentration of 50-200 nM.
Specifications
Specifications
| Numero di accesso | Q13564.1(APPBP1) Q8TBC4.2 (UBA3) |
| Da utilizzare con (applicazione) | Enzyme Activity |
| Formulazione | Supplied as a 0.2 μm filtered solution in HEPES, NaCl, and DTT. |
| ID gene (immissione) | 8883 |
| Peso molecolare | M.W.-theoretical: 62 kDa (APPBP1) & 52 kDa (UBA3), M.W.-observed: 57-63 kDa (APPBP1) & 46-54 kDa (UBA3), under reducing conditions |
| Quantità | 25 μg |
| Sorgente | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human APPBP1/UBA3 Complex protein Ala2-Leu534 with an N-terminal Met and 6-His tag (APPBP1), Met1-Ser463 (UBA3) |
| Requisiti di stoccaggio | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
| Concentrazione di endotossine | <0.10 EU per 1 μg of the protein by the LAL method. |
| Alias gene | A-116A10.1, amyloid beta precursor protein binding protein 1, amyloid beta precursor protein binding protein 1, 59kDa, Amyloid beta precursor protein-binding protein 1, 59 kDa, amyloid beta precursor protein-binding protein 1, 59kD, Amyloid protein-binding protein 1, APPBP1APP-BP1, NAE1, NEDD8 activating enzyme E1 subunit 1, NEDD8-activating enzyme E1 regulatory subunit, NEDD8-activating enzyme E1 subunit, protooncogene protein 1, Proto-oncogene protein 1, UBE1C, ula-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?