missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aquaporin-4 Protein
A DNA sequence encoding the Homo sapiens (Human) Aquaporin-4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5672-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001641.1 | |
Aquaporin-4 Protein | |
Research Use Only | |
AQP4 | |
Recombinant Protein | |
Yeast | |
CPDVEFKRRF KEAFSKAAQQ TKGSYMEVED NRSQVETDDL ILKPGVVHVI DVDRGEEKKG KDQSGEVLSS V | |
Greater than 81% as determined by SDS-PAGE. |
361 | |
10 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |