missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARF3 / ADP-ribosylation factor 3 Protein
A DNA sequence encoding the Homo sapiens (Human) ARF3 / ADP-ribosylation factor 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5673-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001650.1 | |
ARF3 / ADP-ribosylation factor 3 Protein | |
Research Use Only | |
ARF3 | |
Recombinant Protein | |
E. coli | |
GNIFGNLLKS LIGKKEMRIL MVGLDAAGKT TILYKLKLGE IVTTIPTIGF NVETVEYKNI SFTVWDVGGQ DKIRPLWRHY FQNTQGLIFV VDSNDRERVN EAREELMRML AEDELRDAVL LVFANKQDLP NAMNAAEITD KLGLHSLRHR NWYIQATCAT SGDGLYEGLD WLANQLKNKK | |
Greater than 80% as determined by SDS-PAGE. |
377 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |