missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASH2L Protein
A DNA sequence encoding the Homo sapiens (Human) ASH2L, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8184-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001098684.1 | |
ASH2L Protein | |
Research Use Only | |
ASH2L | |
Recombinant Protein | |
E. coli | |
MDTQAGSVDE ENGRQLGEVE LQCGICTKWF TADTFGIDTS SCLPFMTNYS FHCNVCHHSG NTYFLRKQAN LKEMCLSALA NLTWQSRTQD EHPKTMFSKD KDIIPFIDKY WECMTTRQRP GKMTWPNNIV KTMSKERDVF LVKEHPDPGS KDPEEDYPKF GLLDQDLSNI GPAYDNQKQS SAVSTSGNLN GGIAAGSSGK GRGAKRKQQD GGTTGTTKKA RSDPLFSAQR LPPHGYPLEH PFNKDGYRYI LAEPDPHAPD PEKLELDCWA GKPIPGDLYR ACLYERVLLA LHDRAPQLKI SDDRLTVVGE KGYSMVRASH GVRKGAWYFE ITVDEMPPDT AARLGWSQPL GNLQAPLGYD KFSYSWRSKK GTKFHQSIGK HYSSGYGQGD VLGFYINLPE DTETAKSLPD TYKDKALIKF KSYLYFEEKD FVDKAEKSLK QTPHSEIIFY KNGVNQGVAY KDIFEGVYFP AISLYKSCTV SINFGPCFKY PPKDLTYRPM SDMGWGAVVE HTLADVLYHV ETEVDGRRSP PWEP | |
Greater than 90% as determined by SDS-PAGE. |
9070 | |
50 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |