missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATF2 Protein
A DNA sequence encoding the Homo sapiens (Human) ATF2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5688-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001243019.1 | |
ATF2 Protein | |
Research Use Only | |
ATF2 | |
Recombinant Protein | |
E. coli | |
MKFKLHVNSA RQYKDLWNMS DDKPFLCTAP GCGQRFTNED HLAVHKHKHE MTLKFGPARN DSVIVADQTP TPTRFLKNCE EVGLFNELAS PFENEFKKAS EDDIKKMPLD LSPLATPIIR SKIEEPSVVE TTHQDSPLPH PESTTSDEKE VPLAQTAQPT SAIVRPASLQ VPNVLLTSSD SSVIIQQAVP SPTSSTVITQ APSSNRPIVP VPGPFPLLLH LPNGQTMPVA IPASITSSNV HVPAAVPLVR PVTMVPSVPG IPGPSSPQPV QSEAKMRLKA ALTQQHPPVT NGDTVKGHGS GLVRTQSEES RPQSLQQPAT STTETPASPA HTTPQTQSTS GRRRRAANED PDEKRRKFLE RNRAAASRCR QKRKVWVQSL EKKAEDLSSL NGQLQSEVTL LRNEVAQLKQ LLLAHKDCPV TAMQKKSGYH TADKDDSSED ISVPSSPHTE AIQHSSVSTS NGVSSTSKAE AVATSVLTQM ADQSTEPALS QIVMAPSSQS QPSGS | |
Greater than 90% as determined by SDS-PAGE. |
1386 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |