missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP synthase subunit alpha, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) ATP synthase subunit alpha, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5693-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001001935.1 | |
ATP synthase subunit alpha, mitochondrial Protein | |
Research Use Only | |
ATP5A1 | |
Recombinant Protein | |
E. coli | |
QKTGTAEMSS ILEERILGAD TSVDLEETGR VLSIGDGIAR VHGLRNVQAE EMVEFSSGLK GMSLNLEPDN VGVVVFGNDK LIKEGDIVKR TGAIVDVPVG EELLGRVVDA LGNAIDGKGP IGSKTRRRVG LKAPGIIPRI SVREPMQTGI KAVDSLVPIG RGQRELIIGD RQTGKTSIAI DTIINQKRFN DGSDEKKKLY CIYVAIGQKR STVAQLVKRL TDADAMKYTI VVSATASDAA PLQYLAPYSG CSMGEYFRDN GKHALIIYDD LSKMS-QAVA YRQMSLLLRR PPGREAYPGD VFYLHSRLLE RAAKMNDAFG GGSLTALPVI ETQAGDVSAY IPTNVISITD GQIFLETELF YKGIRPAINV GLSVSRVGSA AQTRAMKQVA GTMKLELAQY REVAAFAQFG SDLDAATQQL LSRGVRLTEL LKQGQYSPMA IEEQVAVIYA GVRGYLDKLE PSKITKFENA FLSHVVSQHQ ALLGTIRADG KISEQSDAKL KEIVTNFLAG FEA | |
Greater than 90% as determined by SDS-PAGE. |
498 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |