missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATP1B3 Protein
A cDNA sequence encoding the ATP1B3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11106-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
ATP1B3 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ATP1B3 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |
Greater than 80.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSTMWVMLQ TLNDEVPKYR DQIPSPGLMV FPKPVTALEY TFSRSDPTSY AGYIEDLKKF LKPYTLEEQK NLTVCPDGAL FEQKGPVYVA CQFPISLLQA CSGMNDPDFG YSQGNPCILV KMNRIIGLKP EGVPRIDCVS KNEDIPNVAV YPHNGMIDLK YFPYYGKKLH VGYLQPLVAV QVSFAPNNTG KEVTVECKID GSANLKSQDD RDKFLGRVMF KITARA |