missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ATPAF2 Protein
A DNA sequence encoding the Homo sapiens (Human) ATPAF2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5700-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_663729.1 | |
ATPAF2 Protein | |
Research Use Only | |
ATPAF2 | |
Recombinant Protein | |
E. coli | |
APPTERKRFY QNVSITQGEG GFEINLDHRK LKTPQAKLFT VPSEALAIAV ATEWDSQQDT IKYYTMHLTT LCNTSLDNPT QRNKDQLIRA AVKFLDTDTI CYRVEEPETL VELQRNEWDP IIEWAEKRYG VEISSSTSIM GPSIPAKTRE VLVSHLASYN TWALQGIEFV AAQLKSMVLT LGLIDLRLTV EQAVLLSRLE EEYQIQKWGN IEWAHDYELQ ELRARTAAGT LFIHLCSEST TVKHKLLKE | |
Greater than 90% as determined by SDS-PAGE. |
91647 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |