missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BCKDHA Protein

A cDNA sequence encoding the BCKDHA was constructed and used to recombinantly synthesize the protein.

137.00€ - 1815.00€

Specifica

Nome BCKDHA Protein
Status giuridico Research Use Only
Concentrazione di endotossine Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross-reattività Human
Visualizza altre specifiche

Products 3
Product Code Brand Quantità Price Quantity & Availability  
Product Code Brand Quantità Price Quantity & Availability  
15916268
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP11137-1UG
1 μg
137.00€
1µg
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
15936268
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP11137-5UG
5 μg
212.00€
5µg
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
15926268
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP11137-50UG
50 μg
1815.00€
50µg
View alternative products
missing translation for 'toSaveUpto' 
»
Spedizione stimata: 21-06-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

Specifications

Specifications

BCKDHA Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The BCKDHA solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 5mM DTT, 30% glycerol and 0.2M NaCl.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSSLDDKPQF PGASAEFIDK LEFIQPNVIS GIPIYRVMDR QGQIINPSED PHLPKEKVLK LYKSMTLLNT MDRILYESQR QGRISFYMTN YGEEGTHVGS AAALDNTDLV FGQYREAGVL MYRDYPLELF MAQCYGNISD LGKGRQMPVH YGCKERHFVT ISSPLATQIP QAVGAAYAAK RANANRVVIC YFGEGAASEG DAHAGFNFAA TLECPIIFFC RNNGYAISTP TSEQYRGDGI AARGPGYGIM SIRVDGNDVF AVYNATKEAR RRAVAENQPF LIEAMTYRIG HHSTSDDSSA YRSVDEVNYW DKQDHPISRL RHYLLSQGWW DEEQEKAWRK QSRRKVMEAF EQAERKPKPN PNLLFSDVYQ EMPAQLRKQQ ESLARHLQTY GEHYPLDHFD K
Greater than 80.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Certificates