missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BLyS / TNFSF13B / BAFF Protein
A DNA sequence encoding the Homo sapiens (Human) BLyS / TNFSF13B / BAFF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8298-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001139117.1 | |
BLyS / TNFSF13B / BAFF Protein | |
Research Use Only | |
TNFSF13B | |
Recombinant Protein | |
E. coli | |
QVAALQGDLA SLRAELQGHH AEKLPAGAGA PKAGLEEAPA VTAGLKIFEP PAPGEGNSSQ NSRNKRAVQG PEETVTQDCL QLIADSETPT IQKGSYTFVP WLLSFKRGSA LEEKENKILV KETGYFFIYG QVLYTDKTYA MGHLIQRKKV HVFGDELSLV TLFRCIQNMP ETLPNNSCYS AGIAKLEEGD ELQLAIPREN AQISLDGDVT FFGALKLL | |
Greater than 90% as determined by SDS-PAGE. |
10673 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |