missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human c-MPL / CD110 / TPOR Protein

A DNA sequence encoding the Homo sapiens (Human) c-MPL / CD110 / TPOR, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6376-ye-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16062860

Quantità Prezzo
1 1350.00€ / 500µg
3 1259.96€ / 500µg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_005364.1
c-MPL / CD110 / TPOR Protein
500 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
4352
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MPL
Recombinant Protein
Yeast
QDVSLLASDS EPLKCFSRTF EDLTCFWDEE EAAPSGTYQL LYAYPREKPR ACPLSSQSMP HFGTRYVCQF PDQEEVRLFF PLHLWVKNVF LNQTRTQRVL FVDSVGLPAP PSIIKAMGGS QPGELQISWE EPAPEISDFL RYELRYGPRD PKNSTGPTVI QLIATETCCP ALQRPHSASA LDQSPCAQPT MPWQDGPKQT SPSREASALT AEGGSCLISG LQPGNSYWLQ LRSEPDGISL GGSWGSWSLP VTVDLPGDAV ALGLQCFTLD LKNVTCQWQQ QDHASSQGFF YHSRARCCPR DRYPIWENCE EEEKTNPGLQ TPQFSRCHFK SRNDSIIHIL VEVTTAPGTV HSYLGSPFWI HQAVRLPTPN LHWREISSGH LELEWQHPSS WAAQETCYQL RYTGEGHQDW KVLEPPLGAR GGTLELRPRS RYRLQLRARL NGPTYQGPWS SWSDPTRVET ATETAW
Videos
Documenti