missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1q Protein
A cDNA sequence encoding the C1q was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11228-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
C1q Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
Lyophilized from 0.01M Na2 EDTA solution containing 0.3M NaCl pH 7.5. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 96.0% as determined by SDS-PAGE. |