missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Casein kinase II subunit beta Protein

A DNA sequence encoding the Homo sapiens (Human) Casein kinase II subunit beta, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8381-ec-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16064321

Quantità Prezzo
1 1180.00€ / 500µg
3 1100.47€ / 500µg
Spedizione stimata
26-10-2020
Aggiungi al carrello


Specifica

Specifica

Casein kinase II subunit beta Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CSNK2B
Recombinant Protein
E. coli
SSSEEVSWIS WFCGLRGNEF FCEVDEDYIQ DKFNLTGLNE QVPHYRQALD MILDLEPDEE LEDNPNQSDL IEQAAEMLYG LIHARYILTN RGIAQMLEKY QQGDFGYCPR VYCENQPMLP IGLSDIPGEA MVKLYCPKCM DVYTPKSSRH HHTDGAYFGT GFPHMLFMVH PEYRPKRPAN QFVPRLYGFK IHPMAYQLQL QAASNFKSPV KTIR
Videos
Documenti