missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL18 / PARC / MIP4 Protein

A DNA sequence encoding the Human CCL18 / PARC / MIP4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10204-ec-250ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943088

Quantità Prezzo
1 1970.00€ / 250µg
3 1839.59€ / 250µg
Spedizione stimata
06-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_002979.1
CCL18 / PARC / MIP4 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
AQVGTNKELC CLVYTSWQIP QKFIVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA
6362
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL18
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration of 1.0-10 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documenti