missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL23 / MIP 3 Protein

A DNA sequence encoding the Human CCL23 / MIP 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10238-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943188

Quantità Prezzo
1 1095.00€ / 100µg
3 1020.98€ / 100µg
Spedizione stimata
04-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_005055.3
CCL23 / MIP 3 Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
RVTKDAETEF MMSKLPLENP VLLDRFHATS ADCCISYTPR SIPCSLLESY FETNSECSKP GVIFLTKKGR RFCANPSDKQ VQVCMRMLKL DTRIKTRKN
6368
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL23
The biological activity determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration of 10-50 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
SDS
Documenti