missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CD40/TNFRSF5 Fc Alexa Fluor™ 488 Protein

Codice prodotto. 18787196
Cambia vista
Click to view available options
Quantità:
20 μg
Dimensione della confezione:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18787196 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18787196 Fornitore R&D Systems™ N. del fornitore AFG1493020
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Fc Chimera His-tag

Measured by flow cytometry for its ability to bind anti-human CD40/TNFRSF5 antibody conjugated beads. The concentration of Recombinant Human CD40/TNFRSF5 Fc Chimera His-tag Alexa Fluor™ 488 (Catalog # AFG1493) that produces 50% of the binding response is 15.0-150 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Numero di accesso P25942-1
Da utilizzare con (applicazione) Bioactivity
Formulazione Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
ID gene (immissione) 958
Peso molecolare M.W.-observed: 58-66 kDa, under reducing conditions., M.W.-theroretical: 47 kDa
Quantità 20 μg
Sorgente Mouse myeloma cell line, NS0-derived human CD40/TNFRSF5 protein Human CD40 (Glu21-Arg193) (C-terminus) Accession # P25942-1 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (N-terminus)
Requisiti di stoccaggio Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Concentrazione di endotossine <1.0 EU / 1 μg of the protein by the LAL method.
Alias gene B cell surface antigen CD40, B-cell surface antigen CD40, Bp50B cell-associated molecule, CD40, CD40 antigen, CD40 molecule, TNF receptor superfamily member 5, CD40 type II isoform, CD40L receptor, CDw40, MGC9013, nerve growth factor receptor-related B-lymphocyte activation molecule, p50, TNFRSF5, TNFRSF5CD40 antigen (TNF receptor superfamily member 5), tumor necrosis factor receptor superfamily member 5, tumor necrosis factor receptor superfamily, member 5
Coniugato Alexa Fluor 488
Linea di prodotti R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Proteine CD40/TNFRSF5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.