missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Coagulation Factor VII / FVII / F7 Protein

A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor VII / FVII / F7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6012-ec-500ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15939359

Quantità Prezzo
1 1715.00€ / 500µg
3 1600.44€ / 500µg
Spedizione stimata
13-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_000122.1
Coagulation Factor VII / FVII / F7 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2155
Greater than 90% as determined by SDS-PAGE.
Research Use Only
F7
Recombinant Protein
E. coli
ANAFLEELRP GSLERECKEE QCSFEEAREI FKDAERTKLF WISYSDGDQC ASSPCQNGGS CKDQLQSYIC FCLPAFEGRN CETHKDDQLI CVNENGGCEQ YCSDHTGTKR SCRCHEGYSL LADGVSCTPT VEYPCGKIPI LEKRNASKPQ GR
Videos
SDS
Documenti