missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cytokeratin 8 Protein
A DNA sequence encoding the Homo sapiens (Human) Cytokeratin 8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6284-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001243222.1 | |
Cytokeratin 8 Protein | |
Research Use Only | |
KRT8 | |
Recombinant Protein | |
E. coli | |
SIRVTQKSYK VSTSGPRAFS SRSYTSGPGS RISSSSFSRV GSSNFRGGLG GGYGGASGMG GITAVTVNQS LLSPLVLEVD PNIQAVRTQE KEQIKTLNNK FASFIDKVRF LEQQNKMLET KWSLLQQQKT ARSNMDNMFE SYINNLRRQL ETLGQEKLKL EAELGNMQGL VEDFKNKYED EINKRTEMEN EFVLIKKDVD EAYMNKVELE SRLEGLTDEI NFLRQLYEEE IRELQSQISD TSVVLSMDNS RSLDMDSIIA EVKAQYEDIA NRSRAEAESM YQIKYEELQS LAGKHGDDLR RTKTEISEMN RNISRLQAEI EGLKGQRASL EAAIADAEQR GELAIKDANA KLSELEAALQ RAKQDMARQL REYQELMNVK LALDIEIATY RKLLEGEESR LESGMQNMSI HTKTTSGYAG GLSSAYGGLT SPGLSYSLGS SFGSGAGSSS FSRTSSSRAV VVKKIETRDG KLVSESSDVL PK | |
Greater than 90% as determined by SDS-PAGE. |
3856 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |