missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DEFB1 / Beta-defensin 1 Protein

A DNA sequence encoding the Human DEFB1 / Beta-defensin 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10429-ec-250ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15943828

Quantità Prezzo
1 1970.00€ / 250µg
3 1839.59€ / 250µg
Spedizione stimata
13-01-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_005209.1
DEFB1 / Beta-defensin 1 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GNFLTGLGHR SDHYNCVSSG GQCLYSACPI FTKIQGTCYR GKAKCCK
1672
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
DEFB1
The biological activity determined by a chemotaxis bioassay using CD34+ dendritic cells is in a concentration range of 100.0-1000.0 ng/ml.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized
Videos
SDS
Documenti