missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Dehydrogenase/reductase SDR family member 7 Protein

A DNA sequence encoding the Homo sapiens (Human) Dehydrogenase/reductase SDR family member 7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8296-ec-100ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16013851

Quantità Prezzo
1 434.28€ / 100µg
6 408.22€ / 100µg
Spedizione stimata
07-06-2021
Aggiungi al carrello


Descrizione

Descrizione

Specifica

Specifica

NP_001309209.1
Dehydrogenase/reductase SDR family member 7 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
51635
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DHRS7
Recombinant Protein
E. coli
DGDLTLLWAE WQGRRPEWEL TDMVVWVTGA SSGIGEELAY QLSKLGVSLV LSARRVHELE RVKRRCLENG NLKEKDILVL PLDLTDTGSH EAATKAVLQE FGRIDILVNN GGMSQRSLCM DTSLDVYRKL IELNYLGTVS LTKCVLPHMI ERKQGKIVTV NSILGIISVP LSIGYCASKH ALRGFFNGLR TELATYPGII VSNICPGPVQ SNIVENSLAG EVTKTIGNNG DQSHKMTTSR CVRLMLISMA NDLKEVWISE QPFLLVTYLW QYMPTWAWWI TNKMGKKRIE NFKSGVDADS SYFKIFKTKH D
Videos
SDS
Documenti