missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DERA Protein
A cDNA sequence encoding the DERA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11659-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
51071 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
DERA | |
Human | |
His | |
The DERA solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 20% glycerol. |
DERA Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSAHNRGTEL DLSWISKIQV NHPAVLRRAE QIQARRTVKK EWQAAWLLKA VTFIDLTTLS GDDTSSNIQR LCYKAKYPIR EDLLKALNMH DKGITTAAVC VYPARVCDAV KALKAAGCNI PVASVAAGFP AGQTHLKTRL EEIRLAVEDG ATEIDVVINR SLVLTGQWEA LYDEIRQFRK ACGEAHLKTI LATGELGTLT NVYKASMIAM MAGSDFIKTS TGKETVNATF PVAIVMLRAI RDFFWKTGNK IGFKPAGGIR SAKDSLAWLS LVKEELGDEW LKPELFRIGA STLLSDIERQ IYHHVTGRYA AYHDLPMS |