missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DHRS2 Protein

A DNA sequence encoding the Homo sapiens (Human) DHRS2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7548-ec-200ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16059320

Quantità Prezzo
1 985.00€ / 200µg
4 919.50€ / 200µg
Spedizione stimata
21-10-2020
Aggiungi al carrello


Specifica

Specifica

NP_005785.1
DHRS2 Protein
200 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
10202
Greater than 80% as determined by SDS-PAGE.
Research Use Only
DHRS2
Recombinant Protein
E. coli
MLSAVARGYQ GWFHPCARLS VRMSSTGIDR KGVLANRVAV VTGSTSGIGF AIARRLARDG AHVVISSRKQ QNVDRAMAKL QGEGLSVAGI VCHVGKAEDR EQLVAKALEH CGGVDFLVCS AGVNPLVGST LGTSEQIWDK ILSVNVKSPA LLLSQLLPYM ENRRGAVILV SSIAAYNPVV ALGVYNVSKT ALLGLTRTLA LELAPKDIRV NCVVPGIIKT DFSKVVRIGF MGMSLSGRTS RNIISCRGLG SQRTVQESCP SCALQMPATS TGRTLRWQAT PLGSERSGGG CVAVVPGPGA
Videos
Documenti