missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dihydropyrimidinase-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Dihydropyrimidinase-related protein 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9750-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001184222.1 | |
Dihydropyrimidinase-related protein 1 | |
Research Use Only | |
DPYSL2 | |
Recombinant Protein | |
Yeast | |
MSYQGKKNIP RITSDRLLIK GGKIVNDDQS FYADIYMEDG LIKQIGENLI VPGGVKTIEA HSRMVIPGGI DVHTRFQMPD QGMTSADDFF QGTKAALAGG TTMIIDHVVP EPGTSLLAAF DQWREWADSK SCCDYSLHVD ISEWHKGIQE EMEALVKDHG VNSFLVYMAF KDRFQLTDCQ IYEVLSVIRD IGAIAQVHAE NGDIIAEEQQ RILDLGITGP EGHVLSRPEE VEAEAVNRAI TIANQTNCPL YITKVMSKSS AEVIAQARKK GTVVYGEPIT ASLGTDGSHY WSKNWAKAAA FVTSPPLSPD PTTPDFLNSL LSCGDLQVTG SAHCTFNTAQ KAVGKDNFTL IPEGTNGTEE RMSVIWDKAV VTGKMDENQF VAVTSTNAAK VFNLYPRKGR IAVGSDADLV IWDPDSVKTI SAKTHNSSLE YNIFEGMECR GSPLVVISQG KIVLEDGTLH VTEGSGRYIP RKPFPDFVYK RIKARSRLAE LRGVPRGLYD GPVCEVSVTP KTVTPASSAK TSPAKQQAPP VRNLHQSGFS LSGAQIDDNI PRRTTQRIVA PPGGRANITS LG | |
Greater than 90% as determined by SDS-PAGE. |
1808 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |