missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DYNC2LI1 Protein
A DNA sequence encoding the Homo sapiens (Human) DYNC2LI1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7924-ec-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001180393.1 | |
DYNC2LI1 Protein | |
Research Use Only | |
DYNC2LI1 | |
Recombinant Protein | |
E. coli | |
MPSETLWEIA KAEVEKRGIN GSEGDGAEIA EKFVFFIGSK NGGKTTIILR CLDRDEPPKP TLALEYTYGR RAKGHNTPKD IAHFWELGGG TSLLDLISIP ITGDTLRTFS LVLVLDLSKP NDLWPTMENL LQATKSHVDK VIMKLGKTNA KAVSEMRQKI WNNMPKDHPQ DHELIDPFPV PLVIIGSKYD VFQDFESEKR KVICKTLRFV AHYYGASLMF TSKSEALLLK IRGVINQLAF GIDKSKSICV DQNKPLFITA GLDSFGQIGS PPVPENDIGK LHAHSPMELW KKVYEKLFPP KSINTLKDIK DPARDPQYAE NEVDEMRIQK DLELEQYKRS SSKSWKQIEL DS | |
Greater than 80% as determined by SDS-PAGE. |
51626 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |