missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endophilin-B1 Protein
A DNA sequence encoding the Homo sapiens (Human) Endophilin-B1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8147-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001193580.1 | |
Endophilin-B1 Protein | |
Research Use Only | |
SH3GLB1 | |
Recombinant Protein | |
E. coli | |
MNIMDFNVKK LAADAGTFLS RAVQFTEEKL GQAEKTELDA HLENLLSKAE CTKIWTEKIM KQTEVLLQPN PNARIEEFVY EKLDRKAPSR INNPELLGQY MIDAGTEFGP GTAYGNALIK CGETQKRIGT ADRELIQTSA LNFLTPLRNF IEGDYKTIAK ERKLLQNKRL DLDAAKTRLK KAKAAETRNS SEQELRITQS EFDRQAEITR LLLEGISSTH AHHLRCLNDF VEAQMTYYAQ CYQYMLDLQK QLGSFPSNYL SNNNQTSVTP VPSVLPNAIG SSAMASTSGL VITSPSNLSD LKECSGSRKA RVLYDYDAAN STELSLLADE VITVFSVVGM DSDWLMGERG NQKGKVPITY LELLN | |
Greater than 80% as determined by SDS-PAGE. |
51100 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |