Learn More
enQuireBio™ Recombinant Human FCER1G Protein
A DNA sequence encoding the Homo sapiens (Human) FCER1G, was expressed in the hosts and tags indicated.
Marca:  enQuireBio™ QP6027-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15939459
Quantità Prezzo
1 234.00€ / 50µg
6 218.51€ / 50µg
Spedizione stimata
04-01-2021
Specifica
NP_004097.1
FCER1G Protein
50 μg
Not determined.
Full Length of Mature Protein
Human
GST
Tris-based buffer, 50% glycerol
2207
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FCER1G
Recombinant Protein
E. coli
LGEPQLCYIL DAILFLYGIV LTLLYCRLKI QVRKAAITSY EKSDGVYTGL STRNQETYET LKHEKPPQ