missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fos-related antigen 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Fos-related antigen 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6053-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_005244.1 | |
Fos-related antigen 2 Protein | |
Research Use Only | |
FOSL2 | |
Recombinant Protein | |
E. coli | |
MYQDYPGNFD TSSRGSSGSP AHAESYSSGG GGQQKFRVDM PGSGSAFIPT INAITTSQDL QWMVQPTVIT SMSNPYPRSH PYSPLPGLAS VPGHMALPRP GVIKTIGTTV GRRRRDEQLS PEEEEKRRIR RERNKLAAAK CRNRRRELTE KLQAETEELE EEKSGLQKEI AELQKEKEKL EFMLVAHGPV CKISPEERRS PPAPGLQPMR SGGGSVGAVV VKQEPLEEDS PSSSSAGLDK AQRSVIKPIS IAGGFYGEEP LHTPIVVTST PAVTPGTSNL VFTYPSVLEQ ESPASPSESC SKAHRRSSSS GDQSSDSLNS PTLLAL | |
Greater than 90% as determined by SDS-PAGE. |
2355 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |