missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fructose-2,6-bisphosphatase TIGAR Protein

A DNA sequence encoding the Human Fructose-2,6-bisphosphatase TIGAR, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10157-ec-5ug

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15942928

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

Specifica

Specifica

NP_065108.1
Fructose-2,6-bisphosphatase TIGAR Protein
Research Use Only
TIGAR
The biological activity determined by its ability to protect U2OS cells from apoptosis induced by hydrogen peroxide is in a concentration range of 0.1-5.0 ug/ml, after pretreating with rHuTIGAR-TAT for 4 hours.
Human
TAT
0.2 m filtered 30 % Acetonitrile, 0.1% TFA, lyophilized
57103
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MARFALTVVR HGETRFNKEK IIQGQGVDEP LSETGFKQAA AAGIFLNNVK FTHAFSSDL MRTKQTMHGI LERSKFCKDM TVKYDSRLRE RKYGVVEGKA LSELRAMAK AAREECPVFT PPGGETLDQV KMRGIDFFEF LCQLILKEAD QKEQFSQGS PSNCLETSLA EIFPLGKNHS SKVNSDSGIP GLAASVLVVS HGAYMRSLF DYFLTDLKCS LPATLSRSEL MSVTPNTGMS LFIINFEEGR EVKPTVQCI CMNLQDHLNG LTETRGGYGR KKRRQ
Greater than 96% as determined by SDS-PAGE and HPLC.
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali