missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Gasdermin-C Protein
A DNA sequence encoding the Homo sapiens (Human) Gasdermin-C, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8195-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_113603.1 | |
Gasdermin-C Protein | |
Research Use Only | |
GSDMC | |
Recombinant Protein | |
E. coli | |
MPSMLERISK NLVKEIGSKD LTPVKYLLSA TKLRQFVILR KKKDSRSSFW EQSDYVPVEF SLNDILEPSS SVLETVVTGP FHFSDIMIQK HKADMGVNVG IEVSVSGEAS VDHGCSLEFQ IVTIPSPNLE DFQKRKLLDP EPSFLKECRR RGDNLYVVTE AVELINNTVL YDSSSVNILG KIALWITYGK GQGQGESLRV KKKALTLQKG MVMAYKRKQL VIKEKAILIS DDDEQRTFQD EYEISEMVGY CAARSEGLLP SFHTISPTLF NASSNDMKLK PELFLTQQFL SGHLPKYEQV HILPVGRIEE PFWQNFKHLQ EEVFQKIKTL AQLSKDVQDV MFYSILAMLR DRGALQDLMN MLELDSSGHL DGPGGAILKK LQQDSNHAWF NPKDPILYLL EAIMVLSDFQ HDLLACSMEK RILLQQQELV RSILEPNFRY PWSIPFTLKP ELLAPLQSEG LAITYGLLEE CGLRMELDNP RSTWDVEAKM PLSALYGTLS LLQQLAEA | |
Greater than 90% as determined by SDS-PAGE. |
56169 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |